Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUP1, Mouse, Clone: 10, Abnova™

Catalog No. 89110629 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-629 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-629 Supplier Abnova Corporation Supplier No. MAB1790
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant TUP1.

Specifications

Antigen TUP1
Applications Western Blot
Classification Monoclonal
Clone 10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant TUP1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene TUP1
Gene Alias AAR1/AER2/AMM1/CRT4/CYC9/FLK1/ROX4/SFL2/UMR7
Gene Symbols TUP1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-200 of Yeast TUP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 850445
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.