Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC5, Mouse, FITC, Clone: 2E7/7, Abnova™

Catalog No. 89109928 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-928 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-928 Supplier Abnova Corporation Supplier No. MAB11794
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of TXNDC5.

This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: ADGEDGQDPHSK

Specifications

Antigen TXNDC5
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2E7/7
Conjugate FITC
Description Mouse monoclonal antibody raised against synthetic peptide of TXNDC5.
Dilution Western Blot (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene TXNDC5
Gene Alias ERP46/EndoPDI/Hcc-2/MGC3178/UNQ364
Gene Symbols TXNDC5
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids of 52-63 of human TXNDC5.
Purification Method Protein G Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81567
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.