Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYK2, Mouse, Clone: 8G8E9, 8G9H5, Abnova™

Catalog No. 89113439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-439 Supplier Abnova Corporation Supplier No. MAB8321
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TYK2.

This gene encodes a member of the tyrosine kinase and, more specifically, the Janus kinases (JAKs) protein families. This protein associates with the cytoplasmic domain of type I and type II cytokine receptors and promulgate cytokine signals by phosphorylating receptor subunits. It is also component of both the type I and type III interferon signaling pathways. As such, it may play a role in anti-viral immunity. A mutation in this gene has been associated with hyperimmunoglobulin E syndrome (HIES) - a primary immunodeficiency characterized by elevated serum immunoglobulin E. [provided by RefSeq

Specifications

Antigen TYK2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8G8E9, 8G9H5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant TYK2.
Dilution Western Blot (1:500-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In ascites
Gene TYK2
Gene Alias JTK1
Gene Symbols TYK2
Host Species Mouse
Immunogen Recombinant protein corresponding to truncated human TYK2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7297
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.