Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYMP, Mouse, Clone: P-GF.44C, Abnova™

Catalog No. 89113122 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-122 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-122 Supplier Abnova Corporation Supplier No. MAB7065

Mouse monoclonal antibody raised against full-length recombinant TYMP.

This gene encodes an angiogenic factor which promotes angiogenesis in vivo and stimulates the in vitro growth of a variety of endothelial cells. It has a highly restricted target cell specificity acting only on endothelial cells. Mutations in this gene have been associated with mitochondrial neurogastrointestinal encephalomyopathy. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq

Specifications

Antigen TYMP
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone P-GF.44C
Conjugate Unconjugated
Description thymidine phosphorylase
Formulation In 10mM PBS, pH 7.4 (0.2% BSA)
Gene TYMP
Gene Alias ECGF1, MNGIE, PDECGF, TP, hPD-ECGF
Gene Symbols TYMP
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human TYMP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1890
Test Specificity Thymidine Phosphorylase (P-GF.44C). By Western blot, this antibody detects a band of 55kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.