Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYRO3, Mouse, Clone: 6D6F10, Abnova™

Catalog No. 89108603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-603 Supplier Abnova Corporation Supplier No. MAB10417
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant TYRO3.

Specifications

Antigen TYRO3
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 6D6F10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant TYRO3.
Dilution ELISA (1:10000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene TYRO3
Gene Alias BYK/Brt/Dtk/FLJ16467/RSE/Sky/Tif
Gene Symbols TYRO3
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 138-321 of human TYRO3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7301
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.