Learn More
UBE2J1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant UBE2J1.
Supplier: Abnova Corporation H00051465A01
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum (ER) and may contribute to quality control ER-associated degradation by the ubiquitin-proteasome system. [provided by RefSeq
Sequence: SPAVKRLMKEAAELKDPTDHYHAQPLEDNLFEWHFTVRGPPDSDFDGGVYHGRIVLPPEYPMKPPSIILLTANGRFEVGKKICLSISGHHPETWQPSWSIRTALLAIIGFSpecifications
UBE2J1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant UBE2J1. | |
UBE2J1 | |
CGI-76/HSPC153/HSPC205/HSU93243/MGC12555/NCUBE1/Ubc6p | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_016021 | |
UBE2J1 | |
UBE2J1 (NP_003329, 9 a.a. ∽ 118 a.a) partial recombinant protein with GST tag. | |
RUO | |
51465 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only