Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2J2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107573 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-573 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-573 Supplier Abnova Corporation Supplier No. H00118424A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant UBE2J2.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined. [provided by RefSeq

Sequence: EKGPTLGSIETSDFTKRQLAVQSLAFNLKDKVFCELFPEVVEEIKQKQKAQDELSSRPQTLPLPDVVPDGETHLVQNGIQLLNGHAPGAVPNLAGLQQANR

Specifications

Antigen UBE2J2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant UBE2J2.
Formulation 50% glycerol
Gene UBE2J2
Gene Accession No. NM_058167
Gene Alias NCUBE2/PRO2121
Gene Symbols UBE2J2
Host Species Mouse
Immunogen UBE2J2 (NP_055881, 124 a.a. ∽ 224 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118424
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.