Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2S, Mouse, Clone: AT2C12, Abnova™

Catalog No. 89113207 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-207 Supplier Abnova Corporation Supplier No. MAB7885
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant UBE2S.

This gene encodes a member of the ubiquitin-conjugating enzyme family. The encoded protein is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme-dependent manner, a characteristic property of ubiquitin carrier proteins. [provided by RefSeq

Specifications

Antigen UBE2S
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone AT2C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant UBE2S.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene UBE2S
Gene Alias E2-EPF/E2EPF/EPF5
Gene Symbols UBE2S
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-222 of human UBE2S.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27338
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.