Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2U (B01P), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89107646 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-646 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-646 Supplier Abnova Corporation Supplier No. H00148581B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human UBE2U protein.

Sequence: MHGRAYLLLHRDFCDLKENNYKGITAKPVSEDMMEWEVEIEGLQNSVWQGLVFQLTIHFTSEYNYAPPVVKFITIPFHPNVDPHTGQPCIDFLDNPEKWNTNYTLSSILLALQVMLSNPVLENPVNLEAARILVKDESLYRTILRLFNRPLQMKDDSQELPKDPRKCIRPIKTTSFSDYYQTWSRIATSKATEYYRTPLLKVPNFIGQYYKWKKMDLQHQKEWNLK

Specifications

Antigen UBE2U
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human UBE2U protein.
Formulation PBS with no preservative; pH 7.4
Gene UBE2U
Gene Accession No. NM_152489.1
Gene Alias MGC35130/RP4-636O23.1
Gene Symbols UBE2U
Host Species Mouse
Immunogen UBE2U (NP_689702.1,1 a.a. ∽ 226 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148581
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.