Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin, Mouse, Clone: 6C11-B3, Abnova™

Catalog No. 89136262 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-262 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-262 Supplier Abnova Corporation Supplier No. MAB6646
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Ubiquitin.

Specifications

Antigen Ubiquitin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6C11-B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Ubiquitin.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Host Species Mouse
Immunogen Native purified bovine Ubiquitin (conjugated with KLH).
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes ∼10KDa corresponding to free ubiquitin
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.