Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin, Mouse, Clone: KM691, Abnova™

Catalog No. 89135438 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-438 Supplier Abnova Corporation Supplier No. MAB3353

Mouse monoclonal antibody raised against recombinant Ubiquitin.

Specifications

Antigen Ubiquitin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone KM691
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Ubiquitin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In ascites (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to human Ubiquitin.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts specifically with human ubiquitin. Does not react with BSA.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.