Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBR2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106579 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-579 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-579 Supplier Abnova Corporation Supplier No. H00023304A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant UBR2.

Proteolysis by the ubiquitin-proteasome system controls the concentration of many regulatory proteins. The selectivity of ubiquitylation is determined by ubiquitin E3 ligases, which recognize the substrate's destabilization signal, or degron. The E3 ligase UBR2 participates in the N-end rule pathway, which targets proteins bearing an N-terminal degron, or N-degron (Kwon et al., 2003 [PubMed 14585983]).[supplied by OMIM

Sequence: MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCG

Specifications

Antigen UBR2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant UBR2.
Formulation 50% glycerol
Gene UBR2
Gene Accession No. NM_015255
Gene Alias C6orf133/DKFZp686C08114/KIAA0349/MGC71112/RP3-392M17.3/bA49A4.1/dJ242G1.1/dJ392M17.3
Gene Symbols UBR2
Host Species Mouse
Immunogen UBR2 (NP_056070, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23304
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.