Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCHL1, Mouse, Clone: 31A3, Abnova™

Catalog No. 89109427 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-427 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-427 Supplier Abnova Corporation Supplier No. MAB11258
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against UCHL1.

The protein encoded by this gene belongs to the peptidase C12 family. This enzyme is a thiol protease that hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. This gene is specifically expressed in the neurons and in cells of the diffuse neuroendocrine system. Mutations in this gene may be associated with Parkinson disease

Specifications

Antigen UCHL1
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 31A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against UCHL1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:250-1:500) Western Blot (1-3 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene UCHL1
Gene Alias PARK5/PGP9.5/Uch-L1
Gene Symbols UCHL1
Host Species Mouse
Immunogen Native UCHL1 from human brain.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7345
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.