Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCP1, Mouse, Clone: 4B7, Abnova™

Catalog No. 89105701 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-701 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-701 Supplier Abnova Corporation Supplier No. H00007350M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant UCP1.

Mitochondrial uncoupling proteins (UCP) are members of the family of mitochondrial anion carrier proteins (MACP). UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is expressed only in brown adipose tissue, a specialized tissue which functions to produce heat. [provided by RefSeq]

Sequence: PVDVVKTRFINSPPGQYKSVPNCAMKVFTNEGPTAF

Specifications

Antigen UCP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant UCP1.
Formulation ascites with no preservative
Gene UCP1
Gene Accession No. NM_021833
Gene Alias SLC25A7/UCP
Gene Symbols UCP1
Host Species Mouse
Immunogen UCP1 (NP_068605,232 a.a. ∽ 267 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7350
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.