Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-UGT1A9, Clone: UGT1A9/1229, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2451630.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
UGT1A9 Monoclonal antibody specifically detects UGT1A9 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
UGT1A9 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
EC 2.4.1.17, GNT1, HLUGP4, LUGP4, polypeptide A9, UDP glucuronosyltransferase 1 family, polypeptide A9, UDP-glucuronosyltransferase 1-9, UDP-glucuronosyltransferase 1A9, UDP-glucuronosyltransferase 1-I, UDPGT, UDPGT 1-9, UGT1, UGT1*9, UGT1.9, UGT1-09, UGT1-9, UGT1AI, UGT1I, UGT-1I | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
It recognizes a protein of about 60kDa, which is identified as human UGT1A9. It does not cross-react with the other UGT1A isoforms including UGT1A7, UGT1A8, and UGT1A10 and shows a high degree of specificity. UGT1A9 is a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This MAb binds to human hepatocytes and the majority of human hepatocellular carcinomas (HCC s). In frozen sections, it stains hepatic cells and may be used as a marker of the liver. This MAb also binds to cell preparations of hepatocellular carcinoma biopsies or cell lines. | |
Store at 4C. | |
IgG2b κ |
Flow Cytometry, Immunofluorescence | |
UGT1A9/1229 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
UGT1A9 | |
A synthetic peptide specific to human UGT1A9 protein. | |
0.2 mg | |
Cancer | |
54600 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction