Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UL94, Mouse, Clone: UL.C1, Abnova™

Catalog No. 89113571 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-571 Supplier Abnova Corporation Supplier No. MAB8762
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of UL94.

Sequence: KLVGKSRKHREYRAV

Specifications

Antigen UL94
Applications Western Blot
Classification Monoclonal
Clone UL.C1
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of UL94.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.05% sodium azide)
Gene UL94
Gene Symbols UL94
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acid 26-40 of cytomegalovirus UL94.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3077496
Test Specificity This antibody recognize specifically CMV UL94 protein.
Target Species Virus
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.