Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC13B, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106334 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-334 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-334 Supplier Abnova Corporation Supplier No. H00010497A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant UNC13B.

This gene is expressed in the kidney cortical epithelial cells and is upregulated by hyperglycemia. The encoded protein shares a high level of similarity to the rat homolog, and contains 3 C2 domains and a diacylglycerol-binding C1 domain. Hyperglycemia increases the levels of diacylglycerol, which has been shown to induce apoptosis in cells transfected with this gene and thus contribute to the renal cell complications of hyperglycemia. Studies in other species also indicate a role for this protein in the priming step of synaptic vesicle exocytosis. [provided by RefSeq

Sequence: SKSNNWAPKYNETFHLLLGNEEGPESYELQICVKDYCFAREDRVLGLAVMPLRDVTAKGSCACWCPLGRKIHMDETGLTILRILSQRSNDEVAREFVKLKSESRSTEEGS

Specifications

Antigen UNC13B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant UNC13B.
Formulation 50% glycerol
Gene UNC13B
Gene Accession No. NM_006377
Gene Alias MGC133279/MGC133280/MUNC13/UNC13/Unc13h2/hmunc13
Gene Symbols UNC13B
Host Species Mouse
Immunogen UNC13B (NP_006368, 1482 a.a. ∽ 1591 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10497
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.