Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNG, Mouse, Clone: k1C12, Abnova™

Catalog No. 89109205 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-205 Supplier Abnova Corporation Supplier No. MAB1099
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant UNG.

This gene encodes one of several uracil-DNA glycosylases. One important function of uracil-DNA glycosylases is to prevent mutagenesis by eliminating uracil from DNA molecules by cleaving the N-glycosylic bond and initiating the base-excision repair (BER) pathway. Uracil bases occur from cytosine deamination or misincorporation of dUMP residues. Alternative promoter usage and splicing of this gene leads to two different isoforms: the mitochondrial UNG1 and the nuclear UNG2. [provided by RefSeq

Specifications

Antigen UNG
Applications ELISA, Western Blot
Classification Monoclonal
Clone k1C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant UNG.
Dilution Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene UNG
Gene Accession No. NM_080911
Gene Alias DGU/DKFZp781L1143/HIGM4/UDG/UNG1/UNG15/UNG2
Gene Symbols UNG
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human UNG.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7374
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.