Learn More
UPP1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant UPP1.
Supplier: Abnova Corporation H00007378A01
Description
The 2 known types of pyrimidine nucleoside phosphorylases, uridine phosphorylase (UP; EC 2.4.2.3) and thymidine phosphorylase (TP; EC 2.4.2.4), in the presence of orthophosphate, catalyze the reversible phosphorolysis of uridine and thymidine or deoxyuridine, respectively, to free bases and ribose-1-phosphate or deoxyribose-1-phosphate. Pyrimidine nucleoside phosphorylases can add ribose or deoxyribose to pyrimidine bases to form nucleosides that can be incorporated into RNA or DNA (Watanabe and Uchida, 1995 [PubMed 7488099]).[supplied by OMIM
Sequence: MAATGANAEKAESHNDCPVRLLNPNIAKMKEDILYHFNLTTSRHNFPALFGDVKFVCVGGSPSRMKAFIRCVGAELGLDCPGRDYPNICAGTSpecifications
UPP1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant UPP1. | |
UPP1 | |
UDRPASE/UP/UPASE/UPP | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003364 | |
UPP1 | |
UPP1 (NP_003355, 1 a.a. ∽ 92 a.a) partial recombinant protein with GST tag. | |
RUO | |
7378 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only