Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vaccinia virus, Mouse, Clone: B208M, Abnova™

Catalog No. 89135877 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-877 Supplier Abnova Corporation Supplier No. MAB4938

Mouse monoclonal antibody raised against vaccinia virus.

Specifications

Antigen Vaccinia virus
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone B208M
Conjugate Unconjugated
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen intact Lister strain., Vaccinia virus
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes free Vaccinia virus and infected cells. Reactive with Lister and NYCBH strains.
Target Species Virus
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.