Learn More
VAMP2, Mouse, Clone: 3E5, Abnova™
Mouse monoclonal antibody raised against partial recombinant VAMP2.
Supplier: Abnova Corporation MAB1097
Description
The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Synaptobrevins/VAMPs, syntaxins, and the 25kDa synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. This gene is thought to participate in neurotransmitter release at a step between docking and fusion. The protein forms a stable complex with syntaxin, synaptosomal-associated protein, 25kDa, and synaptotagmin. It also forms a distinct complex with synaptophysin. It is a likely candidate gene for familial infantile myasthenia (FIMG) because of its map location and because it encodes a synaptic vesicle protein of the type that has been implicated in the pathogenesis of FIMG. [provided by RefSeq
Specifications
VAMP2 | |
Monoclonal | |
Unconjugated | |
Western Blot (1:2000-1:3000) The optimal working dilution should be determined by the end user. | |
VAMP2 | |
FLJ11460/SYB2/VAMP-2 | |
Mouse | |
100 μL | |
Primary | |
Human, Rat | |
Liquid |
ELISA, Western Blot | |
3E5 | |
Mouse monoclonal antibody raised against partial recombinant VAMP2. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
NM_014232 | |
VAMP2 | |
Recombinant protein corresponding to amino acids 1-89 of human VAMP2. | |
RUO | |
6844 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.