Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vcam1, Mouse, Clone: eBioMR106, Abnova™

Catalog No. 89109198 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-198 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-198 Supplier Abnova Corporation Supplier No. MAB10983

Mouse monoclonal antibody raised against Vcam1.

Specifications

Antigen Vcam1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone eBioMR106
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Vcam1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Vcam1
Gene Alias MGC108734/VCAM1B
Gene Symbols Vcam1
Host Species Mouse
Immunogen Rat Vcam1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25361
Test Specificity The monoclonal antibody reacts with rat Vcam1, a 110 kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells.
Target Species Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.