Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM1, Mouse, PE, Clone: STA, Abnova™

Catalog No. 89113791 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-791 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-791 Supplier Abnova Corporation Supplier No. MAB9799
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native VCAM1.

This gene is a member of the Ig superfamily and encodes a cell surface sialoglycoprotein expressed by cytokine-activated endothelium. This type I membrane protein mediates leukocyte-endothelial cell adhesion and signal transduction, and may play a role in the development of artherosclerosis and rheumatoid arthritis. Two alternatively spliced transcripts encoding different isoforms have been described for this gene. [provided by RefSeq

Specifications

Antigen VCAM1
Applications Flow Cytometry
Classification Monoclonal
Clone STA
Conjugate PE
Description Mouse monoclonal antibody raised against native VCAM1.
Dilution Flow Cytometry (20 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide)
Gene VCAM1
Gene Alias CD106/DKFZp779G2333/INCAM-100/MGC99561
Gene Symbols VCAM1
Host Species Mouse
Immunogen Native purified VCAM1 from human DS6 T cell line.
Purification Method Size-exclusion chromatography purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7412
Test Specificity The mouse monoclonal antibody STA recognizes VCAM1 (CD106 antigen), a 100-110 kDa type I membrane protein of the immunoglobulin superfamily, a crucial mediator of leukocyte adhesion, and a costimulation molecule.
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.