Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-VEGF R1/Flt-1, Clone: FLT1/658, Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | VEGFR1/Flt-1 |
|---|---|
| Clone | FLT1/658 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1-5ug/ml for coating, Functional |
| Applications | ELISA, Functional Assay |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23461802
![]() |
Novus Biologicals
NBP2346180.2 MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
VEGFR1/Flt-1 Monoclonal antibody specifically detects VEGFR1/Flt-1 in Human samples. It is validated for ELISA, Functional.Specifications
| VEGFR1/Flt-1 | |
| 1.0 mg/mL | |
| ELISA, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases | |
| 10mM PBS - BSA free with No Preservative | |
| EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 | |
| FLT1 | |
| IgG1 κ | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| FLT1/658 | |
| ELISA 1-5ug/ml for coating, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P17948 | |
| 2321 | |
| Recombinant human VEGF-R1 protein | |
| Primary | |
| Protein A purified | |
| Three cell membrane receptor tyrosine kinases, Flt-1 (also designated VEGF-R1), Flk-1 (also designated VEGF-R2) and Flt-4, putatively involved in the growth of endothelial cells, are characterized by the presence of seven immunoglobulin-like sequences in their extracellular domain. These receptors exhibit high degrees of sequence relatedness to each other as well as lesser degrees of relatedness to the class III receptors including CSF-1/Fms, PDGR, SLFR/Kit and Flt-3/Flk-2. Two members of this receptor class, Flt-1 and Flk-1, have been shown to represent high affinity receptors for vascular endothelial growth factors (VEGFs). On the basis of structural similarity to Flt-1 and Flk-1, it has been speculated that Flt-4 might represent a third receptor for either VEGF or a VEGF-related ligand. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title