Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIL1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105720 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-720 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-720 Supplier Abnova Corporation Supplier No. H00007429A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant VIL1.

This gene encodes a member of a family of calcium-regulated actin-binding proteins. This protein represents a dominant part of the brush border cytoskeleton which functions in the capping, severing, and bundling of actin filaments. Two mRNAs of 2.7kb and 3.5kb have been observed; they result from utilization of alternate poly-adenylation signals present in the terminal exon. [provided by RefSeq

Sequence: MTKLSAQVKGSLNITTPGLQIWRIEAMQMVPVPSSTFGSFFDGDCYIILAIHKTASSLSYDIHYWIGQDSSLDEQGA

Specifications

Antigen VIL1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant VIL1.
Formulation 50% glycerol
Gene VIL1
Gene Accession No. NM_007127
Gene Alias D2S1471/VIL
Gene Symbols VIL1
Host Species Mouse
Immunogen VIL1 (NP_009058, 1 a.a. ∽ 77 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7429
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.