Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIM, Mouse, Clone: LN6, Abnova™

Catalog No. 89109630 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-630 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-630 Supplier Abnova Corporation Supplier No. MAB11483
Add to Cart
Add to Cart

mouse monoclonal antibody raised against VIM.

This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a dominant, pulverulent cataract

Specifications

Antigen VIM
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone LN6
Conjugate Unconjugated
Description mouse monoclonal antibody raised against VIM.
Dilution Immunofluorescence (0.5-1.0 ug/mL) Immunohistochemistry (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Culture supernatant with 0.05% Azide
Gene VIM
Gene Alias FLJ36605
Gene Symbols VIM
Host Species Mouse
Immunogen Human thymic nuclear extract.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Bovine, Feline, Human, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.