Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIM, Mouse, Clone: VI-01, Abnova™
Mouse monoclonal antibody raised against native VIM.
Supplier: Abnova Corporation MAB3880
Specifications
| VIM | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against native VIM. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| VIM | |
| Native purified VIM from porcine brain cold stable proteins after depolymerization of microtubules. | |
| RUO | |
| 407747 | |
| Mammalia, Pig | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| VI-01 | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| VIM | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with vimentin, a 57 KDa intermediate filament expressed in varietyof mesenchymal and mesodermal cell types. Cross-reactivity was found with smooth muscledesmin. | |
| Store at 4°C. Do not freeze. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction