Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Vinculin (Ascites Free), Clone: VIIF9 (7F9),

Catalog No. MAB3574C Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3574C 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3574C Supplier MilliporeSigma™ Supplier No. MAB3574C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Vinculin (Ascites Free) clone: VIIF9 (7F9) in Human, Mouse, Rabbit, Rat samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting

Metavinculin (MV) and its a.a. 916-983 truncated splice variant Vinculin (UniProt P18206) are cytoskeleton proteins encoded by the VCL (also known as MVCL, CMD1W, CMH15) gene (Gene ID 7414) in human. The N-terminal globular head domain (a.a. 2-835) of Vinculin/VM contains binding sites for talin and α-actinin as well as a tyrosine phosphorylation site, while their C-terminal Tail region (a.a. 879-1134 of VM; a.a. 879-1066 of Vinculin) contains binding sites for F-actin, paxillin, and lipids. Vinculin is associated with focal adhesion and adherens junctions, which are complexes that nucleate actin filaments and crosslinkers between the external medium, plasma membrane, and actin cytoskeleton. The loss of vinculin disrupts focal adhesion complex formation and prevents cell adhesion and spreading, accompanied by reduced stress fiber formation and inhibition of lamellipodia extension. Smooth muscles and skeletal muscles in their well-differentiated, contractile state co-express vinculin and meta-vinculin. Both vinculin isoforms co-localize in muscular adhesive structures, such as dense plaques in smooth muscles, intercalated discs in cardiomyocytes, and costameres in skeletal muscles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vinculin (Ascites Free)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VIIF9 (7F9)
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P18206
Gene Symbols VCL; MVCL; CMD1W; CMH15
Host Species Mouse
Immunogen Human uterus meta-vinculin.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_003364
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.