Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG1, Mouse, Biotin, Clone: CT1, Abnova™

Catalog No. 89112489 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-489 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-489 Supplier Abnova Corporation Supplier No. MAB5857
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native VSIG1.

Specifications

Antigen VSIG1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone CT1
Conjugate Biotin
Description Mouse monoclonal antibody raised against native VSIG1.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene VSIG1
Gene Alias CHT1
Gene Symbols VSIG1
Host Species Mouse
Immunogen Native purified from chicken thymocytes.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414795
Test Specificity ChT1 antigen (Mr 63/104 KDa) on chicken thymocytes.
Target Species Chicken
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.