Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VTN, Mouse, Clone: BE10, Abnova™

Catalog No. 89112172 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-172 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-172 Supplier Abnova Corporation Supplier No. MAB5220
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native VTN.

The protein encoded by this gene is a member of the pexin family. It is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. It is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond. [provided by RefSeq

Specifications

Antigen VTN
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone BE10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native VTN.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (1% BSA, 0.09% sodium azide)
Gene VTN
Gene Alias V75/VN/VNT
Gene Symbols VTN
Host Species Mouse
Immunogen Native purified human VTN.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7448
Test Specificity This antibody is specific to vitronectin in human tissues. No crossreaction with other matrix molecules. Vitronectin, also known as serum spreading factor, S-protein and epibolin, is a glycoprotein present both in human plasma and serum. Clone BE10 recognizes vitronectin specifically and can be used in immunohistochemical stainings, immunoblottings and other immunoassays for cellular and tumor biology.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.