Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWF, Mouse, Clone: VWF635, Abnova™

Catalog No. 89109519 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-519 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-519 Supplier Abnova Corporation Supplier No. MAB11356
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant VWF.

The glycoprotein encoded by this gene functions as both an antihemophilic factor carrier and a platelet-vessel wall mediator in the blood coagulation system. It is crucial to the hemostasis process. Mutations in this gene or deficiencies in this protein result in von Willebrand's disease. An unprocessed pseudogene has been found on chromosome 22. [provided by RefSeq]

Specifications

Antigen VWF
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone VWF635
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant VWF.
Dilution Immunofluorescence (1:100-1:200) Immunohistochemistry (1:100-1:200 for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein) Western Blot (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Gene VWF
Gene Alias F8VWF/VWD
Gene Symbols VWF
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 845-949 of human VWF.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.