Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WBSCR27, Mouse, Clone: 2A12, Abnova™

Catalog No. 89107659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-659 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-659 Supplier Abnova Corporation Supplier No. H00155368M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant WBSCR27.

This gene encodes a protein belonging to ubiE/COQ5 methyltransferase family. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.22-q11.23. [provided by RefSeq

Sequence: MAQEEGGSLPEVRARVRAAHGIPDLAQKLHFYDRWAPDYDQDVATLLYRAPRLAVDCLTQALPGPPHSALILDVACGTGLVAAELRAPGFLQLHGVDGSPGMLEQARAPGLYQRLSLCTLGQEPLPSPEGTFDAVLIVGALSDGQVPCNAIPELHVTKPGGLVCLTTRTNWSNLQYKEALEATLDRLEQAGMWEGLVAWPVDRLWTAGSWLPPSWWWYPASLPRMASSPALSTCTESGRRPRLRK

Specifications

Antigen WBSCR27
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant WBSCR27.
Formulation PBS with no preservative; pH 7.4
Gene WBSCR27
Gene Accession No. BC030295.2
Gene Alias MGC40131
Gene Symbols WBSCR27
Host Species Mouse
Immunogen WBSCR27 (AAH30295.1,1 a.a. ∽ 245 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155368
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.