Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIPI1, Mouse, Clone: 2D1, Abnova™

Catalog No. 89107005 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-005 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-005 Supplier Abnova Corporation Supplier No. H00055062M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant WIPI1.

WD40 repeat proteins are key components of many essential biologic functions. They regulate the assembly of multiprotein complexes by presenting a beta-propeller platform for simultaneous and reversible protein-protein interactions. Members of the WIPI subfamily of WD40 repeat proteins, such as WIPI1, have a 7-bladed propeller structure and contain a conserved motif for interaction with phospholipids (Proikas-Cezanne et al., 2004 [PubMed 15602573]).[supplied by OMIM

Sequence: GGECVLIKTHSLLGSGTTEENKENDLRPSLPQSYAATVARPSASSASTVPGYSEDGGALRGEVIPEHEFATGPVCLDDENEFPPIILCRGNQKGKTKQ

Specifications

Antigen WIPI1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant WIPI1.
Formulation ascites with no preservative
Gene WIPI1
Gene Accession No. NM_017983
Gene Alias ATG18/FLJ10055/WIPI49
Gene Symbols WIPI1
Host Species Mouse
Immunogen WIPI1 (NP_060453.2,348 a.a. ∽ 445 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55062
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.