Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNK3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107306 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-306 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-306 Supplier Abnova Corporation Supplier No. H00065267A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant WNK3.

Members of the 'with no lysine' (WNK) kinase family, such as WNK3, are serine-threonine protein kinases that lack the almost invariant catalytic lysine in subdomain II, which is important for binding ATP in the catalytic site. Instead, these kinases have a conserved lysine in subdomain I that is thought to provide this function (Holden et al., 2004 [PubMed 15194194]).[supplied by OMIM

Sequence: MATDSGDPASTEDSEKPDGISFENRVPQVAATLTVEARLKEKNSTFSASGETVERKRFFRKSVEMTEDDKVAESSPKDERIKAAMNIPRVDKLPSNVLRG

Specifications

Antigen WNK3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant WNK3.
Formulation 50% glycerol
Gene WNK3
Gene Accession No. NM_020922
Gene Alias KIAA1566/PRKWNK3
Gene Symbols WNK3
Host Species Mouse
Immunogen WNK3 (NP_065973, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65267
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.