Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xgsk-3, Mouse, Clone: G3, Abnova™

Catalog No. 89111582 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-582 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-582 Supplier Abnova Corporation Supplier No. MAB3755
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant xgsk-3.

Specifications

Antigen Xgsk-3
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone G3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant xgsk-3.
Dilution ELISA (1:1000) Immunoprecipiation (5-10 ug) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.01% sodium azide)
Gene Xgsk-3
Gene Alias Gsk3b/MGC131076
Gene Symbols Xgsk-3
Host Species Mouse
Immunogen Recombinant protein corresponding to Xenopus laevis xgsk-3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399097
Test Specificity Recognizes GSK3beta.
Target Species Frog, Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.