Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPO5, Mouse, Clone: 2C5-1B3, Abnova™

Catalog No. 89107195 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-195 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-195 Supplier Abnova Corporation Supplier No. H00057510M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant XPO5.

Exportin-5 belongs to a large family of karyopherins (see MIM 602738) that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.[supplied by OMIM

Sequence: MRLSQKWQVINQRSLLCGEDEAADENPESQEMLEEQLVRMLTREVMDLITVCCVSKKGADHSSAPPADGDDEEMMATEVTPSAMAELTDLGKCLMKHEDVCTALLITAFNSLAWKDTLSCQRTTSQLCWPLLKQVLSGTLLADAVTWLFTSVLKGLQMHGQHDGCMASLVHLAFQIYEALRPRYLEIRAVMEQIPEIQKDSLDQFDCKLLNPSLQKVADKRRKDQFKRLIAGCIGKPLGEQFRKEVHIKNLPSLFKKTKPMLETEVLDNDGGGLATIFEP

Specifications

Antigen XPO5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C5-1B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant XPO5.
Formulation ascites with no preservative
Gene XPO5
Gene Accession No. BC000129
Gene Alias FLJ14239/FLJ32057/FLJ45606/KIAA1291
Gene Symbols XPO5
Host Species Mouse
Immunogen XPO5 (AAH00129,1 a.a. ∽ 280 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57510
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.