Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC5/XRCC6, Mouse, Clone: 162, Abnova™

Catalog No. 89111391 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-391 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-391 Supplier Abnova Corporation Supplier No. MAB3395

Mouse monoclonal antibody raised against native XRCC5/XRCC6.

Specifications

Antigen XRCC5/XRCC6
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 162
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native XRCC5/XRCC6.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene Alias CTC75/CTCBF/G22P1/KU70/ML8/TLAA
Gene Symbols XRCC6
Host Species Mouse
Immunogen Native purified XRCC5/XRCC6 from human B cell nuclei from plasmacytoid 2p68 cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2547/7520
Test Specificity This antibody is known to react with human, monkey and Xenopus. Mouse and rat (immuno-precipitation only). Does not react with bovine and rabbit.
Target Species Frog, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.