Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YDJ1, Mouse, Clone: 1G10.H8, Abnova™

Catalog No. 89110886 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-886 Supplier Abnova Corporation Supplier No. MAB2200
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length YDJ1.

Specifications

Antigen YDJ1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1G10.H8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length YDJ1.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene YDJ1
Gene Alias HSP40/MAS5
Gene Symbols YDJ1
Host Species Mouse
Immunogen Full length yeast YDJ1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 855661
Test Specificity This antibody recognizes yeast Hsp40 (Ydj1). It does not cross-react with human, mouse, or rat Hsp40.
Target Species Yeast
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.