Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YWHAG, Mouse, Clone: HS23, Abnova™

Catalog No. 89110906 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-906 Supplier Abnova Corporation Supplier No. MAB2288
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against YWHAG.

This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the rat ortholog. It is induced by growth factors in human vascular smooth muscle cells, and is also highly expressed in skeletal and heart muscles, suggesting an important role for this protein in muscle tissue. It has been shown to interact with RAF1 and protein kinase C, proteins involved in various signal transduction pathways. [provided by RefSeq

Specifications

Antigen YWHAG
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone HS23
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against YWHAG.
Dilution Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.02% sodium azide)
Gene YWHAG
Gene Alias 14-3-3GAMMA
Gene Symbols YWHAG
Host Species Mouse
Immunogen N-terminus of human YWHAG.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7532
Test Specificity This antibody is specific to human 14-3-3 gamma.
Target Species Bovine, Chicken, Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.