Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP70, Mouse, AP, Clone: SBZAP, Abnova™

Catalog No. 89112521 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-521 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-521 Supplier Abnova Corporation Supplier No. MAB5889
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ZAP70.

This gene encodes an enzyme belonging to the protein tyrosine kinase family, and it plays a role in T-cell development and lymphocyte activation. This enzyme, which is phosphorylated on tyrosine residues upon T-cell antigen receptor (TCR) stimulation, functions in the initial step of TCR-mediated signal transduction in combination with the Src family kinases, Lck and Fyn. This enzyme is also essential for thymocyte development. Mutations in this gene cause selective T-cell defect, a severe combined immunodeficiency disease characterized by a selective absence of CD8-positive T-cells. Two transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen ZAP70
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone SBZAP
Concentration Lot specific
Conjugate Alkaline Phosphatase (AP)
Description Mouse monoclonal antibody raised against partial recombinant ZAP70.
Dilution ELISA (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 50mM Tris, pH 8.0 (1mM MgCl2, 50% glycerol, 0.09% sodium azide)
Gene ZAP70
Gene Alias FLJ17670/FLJ17679/SRK/STD/TZK/ZAP-70
Gene Symbols ZAP70
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 280-309 of human ZAP70.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7535
Test Specificity human TCR ζ; Zap-70 (Mr 70 KDa).
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.