Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB8OS, Mouse, Clone: 1G4, Abnova™

Catalog No. 89107764 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-764 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-764 Supplier Abnova Corporation Supplier No. H00339487M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ZBTB8OS.

Sequence: MAQEEEDVRDYNLTEEQKAIKAKYPPVNRKYEYLDHTADVQLHAWGDTLEEAFEQCAMAMFGYMTDTGTVEPLQTVEVETQGDDLQSLLFHFLDEWLYKFSADEFFIPRVGRRIFIVQAPSGNRSQSNNIFSNAGL

Specifications

Antigen ZBTB8OS
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant ZBTB8OS.
Formulation PBS with no preservative; pH 7.4
Gene ZBTB8OS
Gene Accession No. BC058843.1
Gene Alias ARCH/ARCH2/MGC62007
Gene Symbols ZBTB8OS
Host Species Mouse
Immunogen ZBTB8OS (AAH58843.1,1 a.a. ∽ 136 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339487
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.