Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZHX2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-526 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-526 Supplier Abnova Corporation Supplier No. H00022882A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ZHX2.

The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family. [provided by RefSeq

Sequence: MEQYQHQPMADDHGYDAVARKATKPMAESPKNGGDVVPQYYKDPKKLCEEDLEKLVTRVKVGSEPAKDCLPAKPSEATSDRSEGSSRDGQGSDENEES

Specifications

Antigen ZHX2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ZHX2.
Formulation 50% glycerol
Gene ZHX2
Gene Accession No. NM_014943
Gene Alias AFR1/KIAA0854/RAF
Gene Symbols ZHX2
Host Species Mouse
Immunogen ZHX2 (NP_055758, 691 a.a. ∽ 788 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22882
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.