Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF181, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107763 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-763 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-763 Supplier Abnova Corporation Supplier No. H00339318A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ZNF181.

Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See MIM 604749 for additional information on zinc finger proteins.[supplied by OMIM

Sequence: FIHRSSLIHHQKIHTGEKPYECRECGKAFCCSSHLTRHQRIHTMEKQYECNKCLKVFSSLSFLVQHQSIHTEEKPFECQKCRKSFNQLESLNMHLRNHIRLKPYECSI

Specifications

Antigen ZNF181
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ZNF181.
Formulation 50% glycerol
Gene ZNF181
Gene Accession No. XM_290835
Gene Alias HHZ181/MGC44316
Gene Symbols ZNF181
Host Species Mouse
Immunogen ZNF181 (XP_290835, 354 a.a. ∽ 461 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339318
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.