Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zp3r, Mouse, Clone: 7H12, Abnova™

Catalog No. 89113458 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-458 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-458 Supplier Abnova Corporation Supplier No. MAB8386
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Zp3r.

Specifications

Antigen Zp3r
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 7H12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Zp3r.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene Zp3r
Gene Alias AV264399/SP56
Gene Symbols Zp3r
Host Species Mouse
Immunogen A mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22789
Test Specificity These antibodies specifically recognize mouse sperm protein sp56. They will react with denatured, nonreduced sp56 but not with denatured, reduced sp56. These antibodies do not cross-react with human sperm.
Target Species Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.