Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN22, Mouse, Clone: 19, Abnova™

Catalog No. 89113889 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-889 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-889 Supplier Abnova Corporation Supplier No. MAB9931
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant ZSCAN22.

Specifications

Antigen ZSCAN22
Applications ELISA, Western Blot
Classification Monoclonal
Clone 19
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant ZSCAN22.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene ZSCAN22
Gene Accession No. NM_181846.2
Gene Alias HKR2/MGC126679/MGC138482/ZNF50
Gene Symbols ZSCAN22
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human ZSCAN22.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342945
Test Specificity It can expression in HepG2 whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.