Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse APLP-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3179
This item is not returnable.
View return policy
Description
APLP-1 Polyclonal specifically detects APLP-1 in Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.Specifications
APLP-1 | |
Polyclonal | |
Unconjugated | |
Q03157 | |
DPP4 | |
Mouse myeloma cell line NS0-derived recombinant mouse APLP-1 Gly37-Arg582 Accession # Q03157 | |
100 μg | |
Primary | |
Detects mouse APLP-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human APLP-1 is observed and less than 1% cross-reactivity with recombinant mouse APLP-2 is observed. | |
Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 25 ug/mL, Immunoprecipitation 25 ug/mL | |
amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLP1, APLPAPLP-1 | |
Goat | |
Affinity Purified | |
RUO | |
333 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction