Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse APLP-1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3179
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3179 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3179 Supplier R&D Systems Supplier No. BAF3179
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

APLP-1 Polyclonal specifically detects APLP-1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen APLP-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q03157
Gene Alias amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLP1, APLPAPLP-1
Gene Symbols DPP4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse APLP-1 Gly37-Arg582 Accession # Q03157
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333
Test Specificity Detects mouse APLP-1 in Western blots. In Western blots, approximately 45% cross-reactivity with recombinant human APLP-1 is observed and less than 1% cross-reactivity with recombinant mouse APLP-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.