Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Artemin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1085SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1085SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1085SP Supplier R&D Systems Supplier No. AF1085SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 6 publications

Artemin Polyclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen Artemin
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.3 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Z0L2
Gene Alias artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin
Gene Symbols ARTN
Host Species Goat
Immunogen E. coli-derived recombinant mouse Artemin Ala112-Gly224 Accession # Q9Z0L2
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9048
Test Specificity Detects mouse Artemin in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human GDNF and recombinant rat GDNF is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.