Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Artemin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB1085
This item is not returnable.
View return policy
Description
Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot.Specifications
Artemin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9Z0L2.1 | |
ARTN | |
E. coli-derived recombinant mouse Artemin Ala112-Gly224 Accession # Q9Z0L2.1 | |
500 μg | |
Primary | |
Detects mouse Artemin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Lefty A, recombinant mouse (rm) ART, rmCripto, rhCripto-1, recombinant drosophila DPP, recombinant rat (rr) GDNF, rhLAP, rmLefty-1, rrMIS, rhPersephin, rmNodal, recombinant amphibian TGF-beta 5, rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, rhTGF-alpha , or rhNeurturin is observed. | |
Mouse | |
Purified |
Western Blot | |
185230 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9048 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction