Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Artemin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10851500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10851500 500 μg
MAB10851SP 25 μg
MAB10851100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB10851500 Supplier R&D Systems Supplier No. MAB10851500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Artemin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 185234
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Z0L2.1
Gene Alias artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin
Gene Symbols ARTN
Host Species Rat
Immunogen E. coli-derived recombinant mouse Artemin Ala112-Gly224 Accession # Q9Z0L2.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9048
Test Specificity Detects mouse Artemin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) AgRP, rmCripto, recombinant human (rh) Cripto-1, recombinant drosophila Dpp, recombinant rat GDNF, rhLAP, rhLatent TGF-beta 1, rhLefty A, rmLefty-1, rmMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant Amphibian TGF-beta 5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.